Assay Details
Gene Symbol: tbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214796.1 | NP_999961.1 | 3 - 4 | 816 | 82 | |
XM_005169810.4 | XP_005169867.1 | 3 - 4 | 841 | 82 | ||
GenBank mRNA | AY256970.1 | - | 3 - 4 | 816 | 82 | |
BC116553.1 | - | 3 - 4 | 643 | 82 | ||
BC164097.1 | - | 3 - 4 | 533 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
