Assay Details
Gene Symbol: tbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214796.1 | NP_999961.1 | 5 - 6 | 1086 | 69 | |
XM_005169810.4 | XP_005169867.1 | 5 - 6 | 1114 | 69 | ||
GenBank mRNA | AY256970.1 | - | 5 - 6 | 1086 | 69 | |
BC116553.1 | - | 5 - 6 | 913 | 69 | ||
BC164097.1 | - | 5 - 6 | 803 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
