Assay Details
Gene Symbol: taf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002548.1 | NP_001002548.1 | 5 - 6 | 583 | 87 | |
XM_005161895.3 | XP_005161952.1 | 5 - 6 | 679 | 87 | ||
XM_021469778.1 | XP_021325453.1 | 5 - 6 | 655 | 87 | ||
GenBank mRNA | BC076258.1 | - | 5 - 6 | 583 | 87 | |
BC154290.1 | - | 5 - 6 | 573 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
