Assay Details
Gene Symbol: gtf2h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128422.1 | NP_001121894.1 | 8 - 9 | 658 | 131 | |
GenBank mRNA | BC076236.1 | - | 8 - 9 | 630 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
