Assay Details
Gene Symbol: mpv17l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002567.1 | NP_001002567.1 | 3 - 4 | 774 | 65 | |
GenBank mRNA | BC076231.1 | - | 3 - 4 | 774 | 65 | |
BC086824.1 | - | 3 - 4 | 753 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
