Assay Details
Gene Symbol: desi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003532.1 | NP_001003532.1 | 2 - 3 | 597 | 105 | |
GenBank mRNA | BC078248.1 | - | 2 - 3 | 597 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
