Assay Details
Gene Symbol: bud31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003860.1 | NP_001003860.1 | 1 - 2 | 177 | 87 | |
GenBank mRNA | AY648727.1 | - | 1 - 2 | 177 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
