Assay Details
Gene Symbol: rdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007333.1 | NP_001007334.1 | 1 - 2 | 238 | 116 | |
GenBank mRNA | BC085554.1 | - | 1 - 2 | 238 | 116 | |
BC164634.1 | - | 1 - 2 | 137 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
