Assay Details
Gene Symbol: rdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007333.1 | NP_001007334.1 | 3 - 4 | 536 | 81 | |
GenBank mRNA | BC085554.1 | - | 3 - 4 | 536 | 81 | |
BC164634.1 | - | 3 - 4 | 435 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
