Assay Details
Gene Symbol: fgf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009561.1 | NP_001009561.1 | 2 - 3 | 320 | 112 | |
GenBank mRNA | AB194699.1 | - | 2 - 3 | 320 | 112 | |
BC162921.1 | - | 2 - 3 | 342 | 112 | ||
BC162923.1 | - | 2 - 3 | 342 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
