Assay Details
Gene Symbol: tex261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024426.1 | NP_001019597.1 | 5 - 6 | 586 | 112 | |
GenBank mRNA | BC095136.1 | - | 5 - 6 | 586 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
