Assay Details
Gene Symbol: slc27a2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025299.1 | NP_001020470.1 | 1 - 2 | 478 | 106 | |
GenBank mRNA | BC068405.1 | - | 1 - 2 | 458 | 106 | |
BC098625.1 | - | 1 - 2 | 478 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
