Assay Details
Gene Symbol: nbas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044807.1 | NP_001038272.1 | 24 - 25 | 2737 | 58 | |
XM_005160696.4 | XP_005160753.1 | 24 - 25 | 2876 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
