Assay Details
Gene Symbol: nbas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044807.1 | NP_001038272.1 | 36 - 37 | 4184 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
