Assay Details
Gene Symbol: nbas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044807.1 | NP_001038272.1 | 9 - 10 | 716 | 102 | |
XM_005160696.4 | XP_005160753.1 | 9 - 10 | 855 | 102 | ||
GenBank mRNA | BC135054.1 | - | 9 - 10 | 736 | 102 | |
BC160609.1 | - | 9 - 10 | 736 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
