Assay Details
Gene Symbol: cacna2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044960.2 | NP_001038425.2 | 16 - 17 | 1519 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
