Assay Details
Gene Symbol: cdc42bpab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045094.1 | NP_001038559.1 | 16 - 17 | 2356 | 69 | |
XM_005160749.4 | XP_005160806.1 | 16 - 17 | 3996 | 69 | ||
XM_005160751.4 | XP_005160808.1 | 16 - 17 | 3996 | 69 | ||
XM_017352506.2 | XP_017207995.1 | 16 - 17 | 3993 | 69 | ||
XM_017352507.2 | XP_017207996.1 | 16 - 17 | 3993 | 69 | ||
XM_021468481.1 | XP_021324156.1 | 17 - 18 | 4017 | 69 | ||
XM_021468482.1 | XP_021324157.1 | 17 - 18 | 4014 | 69 | ||
XM_021468483.1 | XP_021324158.1 | 17 - 18 | 4017 | 69 | ||
XM_021468485.1 | XP_021324160.1 | 17 - 18 | 4017 | 69 | ||
XM_021468486.1 | XP_021324161.1 | 17 - 18 | 4017 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
