Assay Details
Gene Symbol: gas2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045098.1 | NP_001038563.1 | 6 - 7 | 685 | 67 | |
XM_005164456.4 | XP_005164513.2 | 6 - 7 | 742 | 67 | ||
XM_021475058.1 | XP_021330733.1 | 6 - 7 | 653 | 67 | ||
GenBank mRNA | BC134975.1 | - | 6 - 7 | 628 | 67 | |
JX013520.1 | - | 5 - 6 | 516 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
