Assay Details
Gene Symbol: ptpdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080596.2 | NP_001074065.2 | 9 - 10 | 2135 | 139 | |
GenBank mRNA | BC129183.1 | - | 9 - 10 | 2117 | 139 | |
BC131875.1 | - | 9 - 10 | 2117 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
