Assay Details
Gene Symbol: rnf217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082853.1 | NP_001076322.1 | 1 - 2 | 956 | 94 | |
GenBank mRNA | BC154340.1 | - | 1 - 2 | 950 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
