Assay Details
Gene Symbol: pim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131539.2 | NP_571614.2 | 2 - 3 | 340 | 90 | |
GenBank mRNA | AF062643.1 | - | 2 - 3 | 314 | 90 | |
BC045836.1 | - | 2 - 3 | 340 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
