Assay Details
Gene Symbol: rplp0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131580.2 | NP_571655.2 | 4 - 5 | 393 | 159 | |
GenBank mRNA | AF134852.1 | - | 4 - 5 | 393 | 159 | |
BC049058.1 | - | 4 - 5 | 405 | 159 | ||
BC062854.1 | - | 4 - 5 | 393 | 159 | ||
BC146738.1 | - | 4 - 5 | 445 | 159 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
