Assay Details
Gene Symbol: u2af1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177479.2 | NP_803432.1 | 6 - 7 | 476 | 110 | |
GenBank mRNA | AF506202.1 | - | 6 - 7 | 476 | 110 | |
BC098601.1 | - | 6 - 7 | 503 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
