Assay Details
Gene Symbol: gria3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198339.1 | NP_938153.1 | 4 - 5 | 726 | 99 | |
XM_005165095.4 | XP_005165152.1 | 4 - 5 | 1192 | 99 | ||
XM_005165096.4 | XP_005165153.1 | 4 - 5 | 1186 | 99 | ||
XM_005165097.4 | XP_005165154.1 | 4 - 5 | 1188 | 99 | ||
XM_005165098.4 | XP_005165155.1 | 4 - 5 | 1190 | 99 | ||
XM_021475991.1 | XP_021331666.1 | 4 - 5 | 1190 | 99 | ||
GenBank mRNA | AF525745.1 | - | 4 - 5 | 726 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
