Assay Details
Gene Symbol: hps4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200326.1 | NP_956620.1 | 12 - 13 | 1996 | 86 | |
GenBank mRNA | BC052117.1 | - | 12 - 13 | 1996 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
