Assay Details
Gene Symbol: atg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200433.2 | NP_956727.1 | 5 - 6 | 523 | 100 | |
XM_005168970.4 | XP_005169027.1 | 5 - 6 | 540 | 100 | ||
GenBank mRNA | BC054904.1 | - | 5 - 6 | 524 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
