Assay Details
Gene Symbol: atg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200433.2 | NP_956727.1 | 6 - 7 | 662 | 71 | |
XM_005168970.4 | XP_005169027.1 | 6 - 7 | 679 | 71 | ||
GenBank mRNA | BC054904.1 | - | 6 - 7 | 663 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
