Assay Details
Gene Symbol: abhd14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207066.2 | NP_996949.2 | 3 - 4 | 517 | 87 | |
GenBank mRNA | BC066291.1 | - | 3 - 4 | 468 | 87 | |
EH433068.1 | - | 3 - 4 | 534 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
