Assay Details
Gene Symbol: eloa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213455.1 | NP_998620.1 | 801 | 71 | ||
GenBank mRNA | BC044429.1 | - | 801 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
