Assay Details
Gene Symbol: gria4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214806.1 | NP_999971.1 | 6 - 7 | 1023 | 71 | |
XM_017359227.2 | XP_017214716.1 | 6 - 7 | 1097 | 71 | ||
XM_017359228.2 | XP_017214717.1 | 6 - 7 | 1102 | 71 | ||
XM_017359229.2 | XP_017214718.1 | 6 - 7 | 1101 | 71 | ||
XM_021481376.1 | XP_021337051.1 | 6 - 7 | 1110 | 71 | ||
GenBank mRNA | AF525747.1 | - | 6 - 7 | 1023 | 71 | |
BC162576.1 | - | 6 - 7 | 951 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
