Assay Details
Gene Symbol: gria4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214806.1 | NP_999971.1 | 7 - 8 | 1193 | 93 | |
XM_017359227.2 | XP_017214716.1 | 7 - 8 | 1267 | 93 | ||
XM_017359228.2 | XP_017214717.1 | 7 - 8 | 1272 | 93 | ||
XM_017359229.2 | XP_017214718.1 | 7 - 8 | 1271 | 93 | ||
XM_021481376.1 | XP_021337051.1 | 7 - 8 | 1280 | 93 | ||
GenBank mRNA | AF525747.1 | - | 7 - 8 | 1193 | 93 | |
BC162576.1 | - | 7 - 8 | 1121 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
