Assay Details
Gene Symbol: arl6ip5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002517.2 | NP_001002517.1 | 1 - 2 | 196 | 69 | |
GenBank mRNA | AB052637.1 | - | 1 - 2 | 178 | 69 | |
BC076281.1 | - | 1 - 2 | 197 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
