Assay Details
Gene Symbol: large2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004538.1 | NP_001004538.1 | 4 - 5 | 687 | 73 | |
XM_017351916.2 | XP_017207405.1 | 5 - 6 | 1251 | 73 | ||
XM_021467635.1 | XP_021323310.1 | 5 - 6 | 654 | 73 | ||
XM_021467636.1 | XP_021323311.1 | 4 - 5 | 810 | 73 | ||
GenBank mRNA | AY662339.1 | - | 4 - 5 | 687 | 73 | |
BC162103.1 | - | 4 - 5 | 639 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
