Assay Details
Gene Symbol: large2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004538.1 | NP_001004538.1 | 6 - 7 | 983 | 76 | |
XM_017351916.2 | XP_017207405.1 | 7 - 8 | 1547 | 76 | ||
XM_021467635.1 | XP_021323310.1 | 7 - 8 | 950 | 76 | ||
XM_021467636.1 | XP_021323311.1 | 6 - 7 | 1106 | 76 | ||
GenBank mRNA | AY662339.1 | - | 6 - 7 | 983 | 76 | |
BC162103.1 | - | 6 - 7 | 935 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
