Assay Details
Gene Symbol: tspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006032.2 | NP_001006032.1 | 2 - 3 | 308 | 101 | |
GenBank mRNA | BC083388.1 | - | 2 - 3 | 279 | 101 | |
BC164666.1 | - | 1 - 2 | 203 | 101 | ||
EH439392.1 | - | 2 - 3 | 309 | 101 | ||
EH569757.1 | - | 2 - 3 | 622 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
