Assay Details
Gene Symbol: slc17a6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009982.1 | NP_001009982.1 | 5 - 6 | 971 | 98 | |
GenBank mRNA | AB183387.1 | - | 5 - 6 | 971 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
