Assay Details
Gene Symbol: hsh2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017541.2 | NP_001017541.2 | 4 - 5 | 673 | 103 | |
GenBank mRNA | BC092186.1 | - | 4 - 5 | 677 | 103 | |
BC164408.1 | - | 3 - 4 | 413 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
