Assay Details
Gene Symbol: asb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017628.1 | NP_001017628.1 | 2 - 3 | 637 | 60 | |
XM_005159719.4 | XP_005159776.1 | 2 - 3 | 1239 | 60 | ||
GenBank mRNA | BC092822.1 | - | 2 - 3 | 637 | 60 | |
BC165810.1 | - | 2 - 3 | 556 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
