Assay Details
Gene Symbol: asb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017628.1 | NP_001017628.1 | 3 - 4 | 1120 | 135 | |
GenBank mRNA | BC092822.1 | - | 3 - 4 | 1120 | 135 | |
BC165810.1 | - | 3 - 4 | 1039 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
