Assay Details
Gene Symbol: ehd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045004.1 | NP_001038469.1 | 1 - 2 | 215 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
