Assay Details
Gene Symbol: c1qtnf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045007.1 | NP_001038472.1 | 1 | 954 | 88 | |
XM_005169953.4 | XP_005170010.1 | 2 | 1127 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
