Assay Details
Gene Symbol: il22ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045279.1 | NP_001038744.1 | 4 - 5 | 563 | 86 | |
GenBank mRNA | BC116595.1 | - | 4 - 5 | 563 | 86 | |
EF014959.1 | - | 4 - 5 | 584 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
