Assay Details
Gene Symbol: il22ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045279.1 | NP_001038744.1 | 5 - 6 | 738 | 88 | |
GenBank mRNA | BC116595.1 | - | 5 - 6 | 738 | 88 | |
EF014959.1 | - | 5 - 6 | 759 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
