Assay Details
Gene Symbol: stra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045312.1 | NP_001038777.1 | 16 - 17 | 1995 | 135 | |
XM_005170689.4 | XP_005170746.1 | 17 - 18 | 1874 | 135 | ||
XM_021470528.1 | XP_021326203.1 | 17 - 18 | 1918 | 135 | ||
GenBank mRNA | BC096850.1 | - | 17 - 18 | 1880 | 135 | |
BC117633.1 | - | 16 - 17 | 1995 | 135 | ||
BC135017.1 | - | 16 - 17 | 2012 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
