Assay Details
Gene Symbol: stra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045312.1 | NP_001038777.1 | 6 - 7 | 886 | 130 | |
XM_005170689.4 | XP_005170746.1 | 7 - 8 | 765 | 130 | ||
XM_021470528.1 | XP_021326203.1 | 7 - 8 | 809 | 130 | ||
GenBank mRNA | BC096850.1 | - | 7 - 8 | 770 | 130 | |
BC117633.1 | - | 6 - 7 | 886 | 130 | ||
BC135017.1 | - | 6 - 7 | 903 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
