Assay Details
Gene Symbol: stra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045312.1 | NP_001038777.1 | 9 - 10 | 1107 | 61 | |
XM_005170689.4 | XP_005170746.1 | 10 - 11 | 986 | 61 | ||
XM_021470528.1 | XP_021326203.1 | 10 - 11 | 1030 | 61 | ||
GenBank mRNA | BC096850.1 | - | 10 - 11 | 992 | 61 | |
BC117633.1 | - | 9 - 10 | 1107 | 61 | ||
BC135017.1 | - | 9 - 10 | 1124 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
