Assay Details
Gene Symbol: cemip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099449.1 | NP_001092919.1 | 5 - 6 | 1395 | 80 | |
XM_005171900.4 | XP_005171957.1 | 6 - 7 | 1633 | 80 | ||
GenBank mRNA | HQ997922.1 | - | 5 - 6 | 1395 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
