Assay Details
Gene Symbol: dvl3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131757.2 | NP_571832.2 | 9 - 10 | 1360 | 62 | |
GenBank mRNA | AB041734.1 | - | 9 - 10 | 1391 | 62 | |
BC044353.1 | - | 9 - 10 | 1342 | 62 | ||
BC098888.1 | - | 9 - 10 | 1333 | 62 | ||
BC165644.1 | - | 9 - 10 | 1026 | 62 | ||
EB922958.1 | - | 3 - 4 | 296 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
