Assay Details
Gene Symbol: glra4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131782.2 | NP_571857.1 | 3 - 4 | 778 | 93 | |
XM_005169598.4 | XP_005169655.1 | 3 - 4 | 709 | 93 | ||
GenBank mRNA | AJ404970.1 | - | 3 - 4 | 782 | 93 | |
BC090676.1 | - | 3 - 4 | 433 | 93 | ||
GQ406229.1 | - | 3 - 4 | 266 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
