Assay Details
Gene Symbol: glra4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131782.2 | NP_571857.1 | 6 - 7 | 1231 | 79 | |
XM_005169598.4 | XP_005169655.1 | 6 - 7 | 1162 | 79 | ||
GenBank mRNA | AJ404970.1 | - | 6 - 7 | 1235 | 79 | |
BC090676.1 | - | 6 - 7 | 886 | 79 | ||
GQ406229.1 | - | 6 - 7 | 719 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
